Learn More
Fursultiamine, TRC
Product Code. 30007600
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007600 50 mg 50mg
30011756 25 mg 25mg
30010999 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007600 Supplier Toronto Research Chemicals Supplier No. TRCF86523550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 804-30-8
Molecular Formula C17 H26 N4 O3 S2
Molecular Weight (g/mol) 398.54
Synonym Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-buten-1-yl]-, Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[(tetrahydrofurfuryl)dithio]-1-butenyl]- (6CI,8CI), Formamide, N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-butenyl]- (9CI), N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[[(tetrahydro-2-furanyl)methyl]dithio]-1-buten-1-yl]formamide, Adventan, Alinamin F, Diteftin, Fursultiamin, Fursultiamine, Judolor, Linamin, N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[4-hydroxy-1-methyl-2-[(tetrahydrofurfuryl)dithio]-1-butenyl]formamide, Retar B1, TTFD, Tetrahydrofurfuryl thiamine disulfide, Thiamin tetrahydrofurfuryl disulfide, Thiamine tetrahydrofurfuryl disulfide
IUPAC Name N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[5-hydroxy-3-(oxolan-2-ylmethyldisulfanyl)pent-2-en-2-yl]formamide
SMILES CC(=C(CCO)SSCC1CCCO1)N(Cc2cnc(C)nc2N)C=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C17H26N4O3S2/c1-12(16(5-6-22)26-25-10-15-4-3-7-24-15)21(11-23)9-14-8-19-13(2)20-17(14)18/h8,11,15,22H,3-7,9-10H2,1-2H3,(H2,18,19,20)
Formula Weight 398.1446
Percent Purity >95%
Chemical Name or Material Fursultiamine
Grade HPLC