Learn More
Furfuryl isopropyl sulfide, 99%, Thermo Scientific™
Catalog Number 11489467
Click to view available options
Quantity:
5 g
25 g
1883-78-9
C8H12OS
156.24
MFCD00040265
WCHRNAKORAINOJ-UHFFFAOYSA-N
furfuryl isopropyl sulfide, 2-isopropylthio methyl furan, isopropyl furfuryl sulfide, furfurylisopropylsulfide, unii-5ott266rl5, fema no. 3161, furan, 2-1-methylethyl thio methyl, furan, 2-isopropylthio methyl, 2-1-methylethyl thio methyl furan
61282
2-(propan-2-ylsulfanylmethyl)furan
CC(C)SCC1=CC=CO1
1883-78-9
C8H12OS
156.24
MFCD00040265
WCHRNAKORAINOJ-UHFFFAOYSA-N
furfuryl isopropyl sulfide, 2-isopropylthio methyl furan, isopropyl furfuryl sulfide, furfurylisopropylsulfide, unii-5ott266rl5, fema no. 3161, furan, 2-1-methylethyl thio methyl, furan, 2-isopropylthio methyl, 2-1-methylethyl thio methyl furan
61282
2-(propan-2-ylsulfanylmethyl)furan
CC(C)SCC1=CC=CO1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Density 1.015
Boiling Point 83°C to 85°C (16 mmHg)
Flash Point 76°C (169°F)
Odor Garlic-like
Refractive Index 1.5055
Quantity 5 g
Beilstein 1306593
Formula Weight 156.25
Percent Purity 99%
Chemical Name or Material Furfuryl isopropyl sulfide