Learn More
Furagin, MedChemExpress
Product Code. p-200100886
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451979 1 g 1g
16400668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16451979 Supplier Medchem Express Supplier No. HY77036
MedChemExpress Furagin, nitrofurantoin analog, is an anti-bacterial agent.
Furagin, nitrofurantoin analog, is an anti-bacterial agent.
Specifications
Chemical Name or Material Furagin
Physical Form Solid
CAS 1672-88-4
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H8N4O5
For Use With (Application) COVID-19-immunoregulation
Synonym Furazidine Furazidin
Solubility Information DMSO : 25 mg/mL (94.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NC(CN1/N=C/C=C/C2=CC=C(O2)[N+]([O-])=O)=O
Molecular Weight (g/mol) 264.19
Formula Weight 264.19
Percent Purity 98.6%
Grade Research