Learn More
Funnel Stand
Product Code. 12926796
Change view
Click to view available options
Material:
Hardwood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12926796 Hardwood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12926796 Supplier S Murray SAMCO Supplier No. L327/01
  • ONLINE EXCLUSIVE PRICE
    481.00 NOK
    402.48 NOK / Pack of 1
    Save 78.52 NOK
    16% Off
Estimated Shipment: 09-12-2026
Hardwood, with interchangeable components
Hardwood, with interchangeable components. Bases are 205mm x 110mm (single) and 290mm x 110mm (double). Rods are 460mm x 16mm (dia). Burette clamps have lined jaws and funnel holders are slotted; all are fitted with a plastic screw and rubber feet on base.
Specifications
Material Hardwood
Diameter (Metric) Opening 460 mm
Product Type Funnel Stand