Learn More
WTW™ Funnel Set
Product Code. 10292401
Change view
Click to view available options
For Use With (Equipment):
CellOx 325 DO Sensor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10292401 CellOx 325 DO Sensor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10292401 Supplier WTW™ Supplier No. 205710
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 10-12-2026
Specifications
Type Funnel Set
For Use With (Application) BOD measurements in NS 19 Winkler bottles
For Use With (Equipment) CellOx 325 DO Sensor