Learn More
Fumaric Acid tert-Butyl Ester (>90%), TRC
Product Code. 30406516
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406516 25 g
30397899 500 mg
30400619 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30406516 Supplier Toronto Research Chemicals Supplier No. TRCF50039525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135355-96-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
Synonym (2E)-2-Butenedioic Acid Mono(1,1-dimethylethyl) Ester, (E)-4-tert-Butoxy-4-oxobut-2-enoic Acid
SMILES CC(C)(C)OC(=O)\C=C\C(=O)O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C8H12O4/c1-8(2,3)12-7(11)5-4-6(9)10/h4-5H,1-3H3,(H,9,10)/b5-4+
Formula Weight 172.0736
Chemical Name or Material Fumaric Acid tert-Butyl Ester