Learn More
Full Range Indicator, pH Range 1-13, for pH Measurement, Fisher Chemical™
Product Code. p-7073649 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
0.1 L
500mL
Product Code. 10366340
Brand: Fisher Chemical U/0025/PB05
Request Bulk or Custom Format
Full Range Indicator, pH Range 1-13, for pH Measurement, Fisher Chemical, Quantity: 500mL, Packaging: Plastic Coated Glass bottle, MDL Number: 283286, Physical Form: Liquid
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 283286
Specifications
Packaging Plastic Coated Glass bottle
Quantity 100 mL
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Full range indicator, pH range 1-13
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol