Learn More
MULTIROIR Full Eurobox Container
Product Code. 17266900| Product Code. | Dimensions (L x W x H) | Color | unitSize |
|---|---|---|---|
| 17266900 | 600 x 400 x 100 mm | White | Each |
| 17236890 | 400 x 300 x 117 mm | Gray | Each |
| 17246890 | 400 x 300 x 220 mm | Gray | Each |
| 17256890 | 400 x 300 x 290 mm | Gray | Each |
| 17266890 | 600 x 400 x 220 mm | Gray | Each |
| 17276890 | 600 x 400 x 320 mm | Gray | Each |
| 17286890 | 600 x 400 x 420 mm | Gray | Each |
| 17296890 | 400 x 300 x 120 mm | Blue | Each |
| 17206900 | 400 x 300 x 120 mm | White | Each |
| 17216900 | 400 x 300 x 120 mm | Red | Each |
| 17226900 | 400 x 300 x 220 mm | Blue | Each |
| 17236900 | 400 x 300 x 220 mm | White | Each |
| 17246900 | 400 x 300 x 220 mm | Red | Each |
| 17256900 | 600 x 400 x 100 mm | Blue | Each |
| 17276900 | 600 x 400 x 100 mm | Red | Each |
| 17286900 | 600 x 400 x 220 mm | Blue | Each |
| 17296900 | 600 x 400 x 220 mm | White | Each |
| 17206910 | 600 x 400 x 220 mm | Red | Each |
| 17216910 | 600 x 400 x 320 mm | Blue | Each |
| 17226910 | 600 x 400 x 320 mm | White | Each |
| 17236910 | 600 x 400 x 320 mm | Red | Each |
Product Code. 17266900
Brand: MULTIROIR BA12018
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
- Full Eurobox container
- The best quality / price ratio on the market
- Solid bottoms and solid walls
- Ideal for the storage and transport of goods
- Stackable and high load capacity
- Temperature resistant from -20 ° C to + 110 ° C
Specifications
Specifications
| Type | Storage Bin |
| Material | Polypropylene |
| Color | White |
| Shape | Rectangular |
| Dimensions (L x W x H) | 600 x 400 x 100 mm |