Learn More
Fukutin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16848344 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16848344 150 μL 150µL
16858344 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16848344 Supplier Proteintech Supplier No. 182761AP150UL
Rabbit Polyclonal Antibody
FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
Specifications
Antigen Fukutin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FKTN
Gene Accession No. O75072, Q8R507
Gene Alias CMD1X, FCMD, FKTN, Fukutin, LGMD2M
Gene Symbols Fktn
Host Species Rabbit
Immunogen Fukutin Fusion Protein Ag13067
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2218, 246179, 362520
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG