missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16420-1-AP-150UL
This item is not returnable.
View return policy
Description
Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids.Specifications
| FUCA1 | |
| Polyclonal | |
| Unconjugated | |
| FUCA1 | |
| Alpha L fucosidase 1, Alpha L fucosidase I, FUCA, FUCA1, fucosidase, alpha L 1, tissue, Tissue alpha L fucosidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24375, 2517, 71665 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04066, P17164, Q99LJ1 | |
| Fuca1 | |
| FUCA1 Fusion Protein Ag9612 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction