missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FTO Monoclonal antibody specifically detects FTO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | FTO |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AG103 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 |
| Host Species | Mouse |
| Immunogen | Recombinant human FTO. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?