Learn More
FTCD Mouse anti-Human, Unconjugated, Clone: 1H8A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66979-1-IG-20UL
 View more versions of this product
Product Code. 16809325
  • 1710.33 NOK / 20µL
Description
The FTCD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FTCD. This antibody recognizes human, mouse, rat, pig antigen. The FTCD antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FTCD
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry
1H8A1
Unconjugated
20 μL
10841
IgG2b