Learn More
FTCD Mouse anti-Human, Unconjugated, Clone: 1H8A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66979-1-IG-20UL
Description
The FTCD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FTCD. This antibody recognizes human, mouse, rat, pig antigen. The FTCD antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| FTCD | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry | |
| 1H8A1 | |
| Unconjugated | |
| 20 μL | |
| 10841 | |
| IgG2b |