missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FSCN1 (Human) Recombinant Protein
Human FSCN1 full-length ORF (NP_003079.1, 1 a.a. - 493 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00006624-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTANGTAEAVQIQFGLINCGNKYLTAEAFGFKVNASASSLKKKQIWTLEQPPDEAGSAAVCLRSHLGRYLAADKDGNVTCEREVPGPDCRFLIVAHDDGRWSLQSEAHRRYFGGTEDRLSCFAQTVSPAEKWSVHIAMHPQVNIYSVTRKRYAHLSARPADEIAVDRDVPWGVDSLITLAFQDQRYSVQTADHRFLRHDGRLVARPEPATGYTLEFRSGKVAFRDCEGRYLAPSGPSGTLKAGKATKVGKDELFALEQSCAQVVLQAANERNVSTRQGMDLSANQDEETDQETFQLEIDRDTKKCAFRTHTGKYWTLTATGGVQSTASSKNASCYFDIEWRDRRITLRASNGKFVTSKKNGQLAASVETAGDSELFLMKLINRPIIVFRGEHGFIGCRKVTGTLDANRSSYDVFQLEFNDGAYNIKDSTGKYWTVGSDSAVTSSGDTPVDFFFEFCDYNKVAIKVGGRYLKGDHAGVLKASAETVDPASLWEY
Specifications
NP_003079.1 | |
fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FSCN1 | |
GST |
6624 | |
Wheat germ expression system | |
25 μg | |
FLJ38511, SNL, p55 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |