Learn More
Macherey-Nagel™ FS-CW 20 M-AM GC Capillary Column
Product Code. 10779942 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
10 m
25 m
50 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.2 μm
0.25 μm
0.35 μm
1 μm
Unit Size:
Each
Pack of 25
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
10779942 50 m 0.32 mm 0.25 μm Pack of 50
10545571 10 m 0.1 mm 0.2 μm Each
10535571 25 m 0.25 mm 0.25 μm Pack of 25
10093131 25 m 0.32 mm 0.25 μm Each
10449561 25 m 0.53 mm 1 μm Each
10184561 50 m 0.25 mm 0.25 μm Each
10358601 50 m 0.32 mm 0.35 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10779942 Supplier Macherey-Nagel™ Supplier No. 733299.50
  • ONLINE EXCLUSIVE PRICE
    14305.00 NOK
    12583.17 NOK / Pack of 50
    Save 1721.83 NOK
    12% Off
Estimated Shipment: 23-11-2026
Specifications
For Use With (Application) Gas Chromatography (GC)
Film Thickness 0.25 μm
Length (Metric) 50 m
Packing Material FS-CW 20 M-AM
Quantity 1 Each
Diameter (Metric) 0.5 mm
USP Type G16
Stationary Phase FS-CW 20 M-AM
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Temperature Range (Metric) 220°C to 240°C