Learn More
OHAUS™ Frontier™ 5707 Multi Centrifuge
Product Code. p-200132130 Shop All Ohaus Products
Click to view available options
Capacity:
4 x 50 mL RB/FA or 4 x 15 mL RB/FA
8 x 15 mL
Unit Size:
1 unit
Each
Product Code. 16241742
Brand: OHAUS™ 30393189
Reliable compact low-speed centrifuge for processing small numbers of blood collection and urine tubes or standard conical tubes. Designed with convenient operation the FC5707 features a uniquely designed splash-proof front panel with Touch-Wheel control on an intuitive interface.
Designed for easy operations and great user experience
  • Easy-to-Use touch-wheel interface for parameter adjustments
  • Convenient interface shows both set and actual parameters
  • One-touch toggling between RPM/RCF, Acc/Dec
  • Double-line LCD display with light blue backlight
  • Audible signal at the end of each run with melody
  • 10 acceleration/deceleration settings
  • 99 memory sets to quickly store and recall commonly used parameters
Safety features enclosed within small footprint
  • Microprocessors control to help prevent accidents and protect both the user and the equipment
  • Rotor imbalance sensor systems stops the unit when it detects an imbalanced load
  • Sturdy mechanical lid with locking system
Flexible rotor selections and kits to cover wide range of general laboratory and diagnostic applications
  • Maintenance-free induction motor produces maximum G-force of 4445 x g
  • Speeds up to 6800 rpm and quick spin for short runs
  • Angle rotor which can handle 8 x 15 mL RB or 4 x 15 mL FA tubes or Angle combi rotor which can handle 4 x 15 mL or 4 x 50 mL RB/FA tubes
TRUSTED_SUSTAINABILITY
Specifications
Capacity 8 x 15 mL
Max. Speed 6,800 rpm
Max. RCF 4,445 x g
Refrigerated No
Control Speed 200 to 6800 rpm
Display LCD with Backlight
Height (English) Exterior 9.3 in.
Height (Metric) Exterior 236 mm
Length (English) Exterior 14 in.
Length (Metric) Exterior 356 mm
Memory 99 Programs
Noise Level ≤60±2 dBA
Safety Features CE, FCC
Width (English) Exterior 10.9 in.
Width (Metric) Exterior 277 in.
Wattage 50 W
Certifications/Compliance CE, FCC
Type Centrifuge
Description FC5707+R05
Dimensions (L x W x H) Exterior 356 x 277 x 236 mm
Electrical Requirements AC Power (Included)
Depth (English) 14 in.
Depth (Metric) 356 mm
Maximum Speed Range 200 to 6800 rpm
Rotor Included Yes
Dimensions (HWD) 236 x 277 x 356 mm
Net Weight 10 kg
Run Time 10 sec. to 99 hr. 59 min. 59 sec., Continous, Quickspin
Product Line Frontier 5000 Series Multi
Voltage 100/230 V
Model FC5707+R05