Learn More
Frizzled 9 Mouse anti-Human, Unconjugated, Clone: 1G9G4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67023-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889395
  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
Description
The Frizzled 9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Frizzled 9. This antibody recognizes human antigen. The Frizzled 9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Frizzled 9
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1G9G4
Unconjugated
20 μL
8326
IgG1