missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24272-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.Specifications
Frizzled 2 | |
Polyclonal | |
Unconjugated | |
FZD2 | |
Frizzled 2, Fz 2, FZD2, FzE2, hFz2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2535, 57265, 64512 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08464, Q14332, Q9JIP6 | |
Fzd2 | |
Frizzled 2 Fusion Protein Ag18003 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |