Learn More
Vileda Professional™ CE Trolley 6L Bucket
Product Code. 17812466
Change view
Click to view available options
Capacity (Metric):
6 L
Unit Size:
Pack of 10
Product Code. Capacity (Metric) unitSize
17812466 6 L Pack of 10
1 options
Product Code. 17812466
Brand: Vileda Professional™ 120942
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Color coding clips.
For use with CE Bucket Trolley or CE Pre-Prepared Trolley
  • Material: Polypropylene
  • Autoclavable
  • 10/Cs.
Specifications
Capacity (Metric) 6 L