Learn More
Labconco FreeZone 18L Console Freeze Dryer
Click to view available options
Material:
PTFE Coil and Collector Chamber
Stainless Steel Coil and Collector Chamber
Includes:
Console
Console Dryer
Dryer, Purge Valve
Purge Valve
Purge Valve, Console
Purge Valve, Mini Chamber
Purge Valve, Mini Chamber, Console Dryer
Purge Valve, Stoppering Tray Dryer
Stoppering Tray Dryer
Unit Size:
1 unit
Description
- Stainless steel or PTFE coil holds 18 liters of ice before defrosting.
- The hp HCFC/CFC-free refrigeration system cools the collector
- Includes a stoppering tray dryer with heated and cooled shelves, vacuum release valve control, stoppering knob and more.
- Lyo-Works™ OS provides a real time display of collector temperature, vacuum level and optional End-Zone™ end point detection system
- Large, intuitive 5 in. full color, capacitive touchscreen display for easy operation.
- Stay Connected
- View Real-Time Sample Data
- Get Consistent, High Quality Results
- Make Challenging Samples Easy
Specifications
Specifications
| Type | Console Freeze Dryer |
| Capacity (Metric) | 18 L |
| No. of Shelves | 3 |
| Temperature Range (Metric) | -50°C |
| Material | Stainless Steel Coil and Collector Chamber |
| Includes | Dryer, Purge Valve |
| Plug Type | All except North America |
| Voltage | 230 V |