Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Click to view available options
Dimensions (LxW):
0.375 x 0.375 in.
0.375 x 0.825 in.
0.5 x 0.5 in.
0.5 x 1 in.
12.70 mm (W) x 41.91 mm (H)
22.86 mm (W) x 12.70 mm (H) x 11.18 mm (Dia)
22.86 mm (W) x 25.40 mm (H)
25 ft. x 0.5 in.
25.40 mm (W) x 12.70 mm (H) x 9.53 mm (Dia)
31.75 mm (W) x 6.35 mm (H)
31.75 mm (W) x 9.53 mm (H)
41.28 mm (W) x 15.24 mm (H) x 9.53 mm (Dia)
44.45 mm (W) x 25.40 mm (H)
For Use With (Equipment):
BMP41, BMP51, M511
BMP51;BMP53;M511;M510
Format:
Continuous
Die-cut
Unit Size:
Each
Pack of 180
Pack of 200
Pack of 220
Pack of 240
Pack of 360
Description
- Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
- Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
- Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | RoHS Compliant to 2011/65/EU (Pass 2015/863) |
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Format | Die-cut |
| Material | Polyester |
| Product Type | Label |
| For Use With (Application) | General Identification, Laboratory Labeling |
| For Use With (Equipment) | BMP41, BMP51, M511 |
| Quantity | 220 Labels/Cartridge |
| Show More |