Learn More
Brady™ FREEZERBONDZ™ Polyester Labels
Product Code. p-8001046
Click to view available options
Size (Metric):
12.7 mm dia.
48.3 W x 25.4 mm H
Unit Size:
Each
Product Code. 15180143
Brand: Brady™ 142940
For use in laboratory identification applications such as vials, centrifuge tubes, test tubes, straws and slides.
  • Polyester
  • Applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Provides excellent print smudge resistance, solvent resistance and low-temperature performance
Specifications
Certifications/Compliance RoHS compliant to 2002/618/EC MCV amendment to RoHS Directive 2011/65/EU
Description FreezerBondz Labels for the BBP33 Printer
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material Polyester
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For laboratory identification
For Use With (Equipment) BBP™ 33
Horizontal Repeat (Metric) 15.24 mm
Labels per Row 3
Min. Operating Temperature (Metric) -196°C
No. per Pack 100/Pk.
Quantity 2500/Pk.
Shape Rectangle with Circle
Size (Metric) 12.7 mm dia.
Software Type LabelMark, MarkWare
Temperature Range (English) -320°F to +266°F
Temperature Range (Metric) -196°C to +130°C
Thickness (English) 0.0026 in.
Thickness (Metric) 0.013 mm
Vertical Repeat (Metric) 15.88 mm
Dimensions (LxW) 12.7 x 12.7 mm
Length (English) 0.5 in.
Width (English) 0.5 in.
Width (Metric) 12.70 mm
Height (Metric) 36.5 mm
Diameter (English) 0.5 in.
Diameter (Metric) 12.7 mm
Alternate Part Number B33-144-490