Learn More
Brady™ FreezerBondz™ Polyester Autoclave and Cryogenic Labels for M710
Product Code. 17837934
Change view
Click to view available options
Dimensions (LxW):
12.7 x 25.4 mm
15.24 x 41.28 mm
25.4 x 25.4 mm
25.4 x 48.26 mm
9.53 x 25.4 mm
Shape:
Rectangle
Rectangle with Circle
Square
Thickness (Metric):
0.065 mm
0.081 mm
Certifications/Compliance:
ASTM D 1000
ASTM D 1000, ASTM G 155
Unit Size:
Pack of 250
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Shape Thickness (Metric) Certifications/Compliance unitSize
17837934 12.7 x 25.4 mm Rectangle 0.081 mm ASTM D 1000 Pack of 500
17857934 9.53 x 25.4 mm Rectangle 0.081 mm ASTM D 1000 Pack of 500
17827984 12.7 x 25.4 mm Rectangle with Circle 0.081 mm ASTM D 1000 Pack of 500
17818024 15.24 x 41.28 mm Rectangle 0.065 mm ASTM D 1000, ASTM G 155 Pack of 500
17828094 25.4 x 25.4 mm Square 0.081 mm ASTM D 1000 Pack of 250
17817944 25.4 x 48.26 mm Rectangle 0.081 mm ASTM D 1000 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17837934 Supplier Brady™ Supplier No. 311982
FreezerBondz™ Label Printer Labels for use with Brady™ Printer M710.
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance, solvent resistance
  • High/low-temperature performance range of -196°C to +100°C
Specifications
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Cryogenic Label
Sterility Non-sterile
For Use With (Application) General Identification, Laboratory Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -196°C
Quantity 500 Labels
Shape Rectangle
Temperature Range (English) -320°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Dimensions (LxW) 12.7 x 25.4 mm
Length (English) 0.5 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 25.4 mm
Alternate Part Number M7-17-492