Learn More
Brady™ FreezerBondz™ Cryogenic Matte Polyester Labels
Click to view available options
Dimensions (LxW):
0.6 x 1.8 in.
1.65 x 0.5 in.
31.75 mm (W) x 9.53 mm (H)
38.10 mm (W) x 55.88 mm (H)
38.10 mm (W) x 95.25 mm (H)
44.45 mm (W) x 25.40 mm (H)
For Use With (Equipment):
BMP41, BMP51, M511
BMP51;BMP53;M511;M510
Unit Size:
Pack of 140
Pack of 180
Pack of 360
Pack of 80
Description
- Designed for laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws and slides
- Withstands solvents, smudges and extreme temperatures, including storage in liquid nitrogen
- Made with a permanent acrylic adhesive for use on flat frozen surfaces or wrapping around tubes and vials with a 1/8 in. overlap
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | 2011/65/EU, 2015/863 |
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Polyester |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| Show More |