Learn More
Eppendorf™ Freezer Rack: CryoCube™ FC660 series, 2 in/53 mm
Product Code. 15713265
Product Code. 15713265 Supplier Eppendorf™ Supplier No. 6001050211
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: CryoCube™ FC660 series, locking rod, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 721 (5.8 x 5.6 x 28.4 in)- Max. box number per rack: 13 (13 x 1)- Stainless steel- For ULT freezer: CryoCube™ FC660/ FC660h (max. 36 racks)- For ULT freezer: CryoCube™ FC660/ FC660h (max. 468 boxes)- With locking rod for increased sample safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack