Learn More
Azpack™ Freeze Drying Stoppers
Product Code. 10483274
Change view
Click to view available options
For Use With (Equipment):
2mL glass vials
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10483274 2mL glass vials Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10483274 Supplier Azpack™ Supplier No. 10483274
  • ONLINE EXCLUSIVE PRICE
    1635.00 NOK
    1397.58 NOK / Pack of 500
    Save 237.42 NOK
    14% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Push in to engage and pull out to remove stopper. Azpack™ Freeze Drying Stoppers are nonthreaded and constructed of sturdy rubber.
  • For use with Freeze Drying and Injection Vials
  • Nonthreaded push-in stopper
  • Use with appropriate polypropylene screw thread cap
Specifications
Material Bromobutyl Rubber
Quantity 500 Pack
For Use With (Equipment) 2mL glass vials
Diameter (Metric) 13 mm