Learn More
Gardner Denver Welch™ Kit 6 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 25 KF
Product Code. 13226349 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Rotary vane pump DN 25 KF
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13226349 Rotary vane pump DN 25 KF Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13226349 Supplier Gardner Denver Welch™ Supplier No. 330045
  • ONLINE EXCLUSIVE PRICE
    2015.00 NOK
    1913.08 NOK / Pack of 1
    Save 101.92 NOK
    5% Off
 Restricted Item
Estimated Shipment: 06-04-2027
For rotary vane pump DN 25 KF
Vacuum concentrator kit includes:
  • KF25 flange adapter
  • 1.5m PVC vacuum hose
  • Clamps and centring rings
TRUSTED_SUSTAINABILITY
Specifications
Type Freeze Dryer and Vacuum Concentrator Adapter Kit
For Use With (Equipment) Rotary vane pump DN 25 KF