missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Free Chlorine Reagent Set for PCA Series Process Analyzers

Product Code. 10729061
Change view
Click to view available options
For Use With (Application):
Short-term Measurements
Unit Size:
Each
Product Code. For Use With (Application) unitSize
10729061 Short-term Measurements Each
1 options
This item is not returnable. View return policy

Product Code. 10729061

Brand: Hanna Instruments™ HI70480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activate the indicator solution before use. Hanna Instruments™ Free Chlorine Reagent Set for PCA Series Process Analyzers are used for chlorine measurement in fresh water. Sets are available for use with short-term and long-term measurements.

Each set includes a 500mL bottle of indicator reagent, a 500mL bottle of buffer reagent and powdered DPD reagent (either in a bottle or individual packets). To use the reagent set, the solution will need to be activated:

  • Add the powder from the bottle or packets into the bottle containing indicator solution
  • Replace the cap and shake the bottle for a few minutes to make sure that the powder is completely dissolved
  • The indicator solution is now activated and ready to be installed in the PCA (Process Color Analyzer)
  • Shake the buffer solution bottle well before installing it in the analyzer—any residual precipitate will not affect readings

Specifications

For Use With (Application) Short-term Measurements
For Use With (Equipment) Hanna PCA Series process analyzers
Includes 500mL Indicator Solution, 500mL Buffer Solution, 5 packets DPD reagent
Quantity 1 Set
Sufficient For Process applications
Test Method DPD
compliance-icons compliance-icons
Product Identifier
  • Reagenzkit Freies Chlor für PCA 300 Chlor-Analysator
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid, N,N-Diethyl-1,4-phenylenediammonium sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.