missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Fraser Broth Base (Dehydrated)

Product Code. p-200112449
Click to view available options
Quantity:
500 g
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 10760714

Brand: Thermo Scientific™ CM0895B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively enrich and isolate Listeria spp. from food and environmental samples with this medium.

Selectively enrich and isolate Listeria spp. from food and environmental samples with Thermo Scientific™ Oxoid™ Fraser Broth (Dehydrated). The medium is intended for the isolation of Listeria as detailed in ISO11290-1:2017 Horizontal method for the detection and enumeration of Listeria monocytogenes and of Listeria spp.

Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. The medium is intended for the isolation of Listeria as detailed in ISO11290-1:2017 Horizontal method for the detection and enumeration of Listeria monocytogenes and of Listeria spp.

  • Selective: the medium is selective for Listeria
  • Easy-to-read: blackening of the medium is presumptive evidence of the presence of Listeria spp.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, IDF, ISO, NMKL.
Description Fraser Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 8.7L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.