missing translation for 'onlineSavingsMsg'
Learn More

FPR Agonist 43, MedChemExpress

Product Code. p-200101911
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483767

Brand: Medchem Express HY19574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist.

FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist.

Specifications

Chemical Name or Material FPR Agonist 43
Physical Form Powder
CAS 903895-98-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H21ClN4O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (324.79 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=C(C(C)C)N(C)N(C2=CC=CC=C2)C1=O)NC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 384.86
Formula Weight 384.86
Percent Purity 97.12%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.