Learn More
FPR Agonist 43, MedChemExpress
Product Code. 16483767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16483767
Brand: Medchem Express HY19574
MedChemExpress FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist.
FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist.
Specifications
Chemical Name or Material FPR Agonist 43
Physical Form Powder
CAS 903895-98-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H21ClN4O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 125 mg/mL (324.79 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=C(C(C)C)N(C)N(C2=CC=CC=C2)C1=O)NC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 384.86
Formula Weight 384.86
Percent Purity 97.12%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.