missing translation for 'onlineSavingsMsg'
Learn More

FPH2, MedChemExpress

Product Code. p-200102142
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475796

Brand: Medchem Express HY12281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FPH2 induces of functional proliferation of primary human hepatocytes and may lead to the development of new therapeutics for liver diseases.

FPH2 induces of functional proliferation of primary human hepatocytes and may lead to the development of new therapeutics for liver diseases.

Specifications

Chemical Name or Material FPH2
CAS 957485-64-2
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClN5O2S
Quantity 100 mg
Synonym BRD-9424
Solubility Information DMSO : 100 mg/mL (282.62 mM; Need ultrasonic)
SMILES O=C(C1=NN(CC)C=C1NC(NC2=CC(Cl)=CC=C2OC)=S)N
Molecular Weight (g/mol) 353.83
Formula Weight 353.83
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.