Learn More
FPH2, MedChemExpress
Product Code. 16475796
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475796 100 mg 100mg
16465058 10 mg 10mg
16463286 25 mg 25mg
16441297 50 mg 50mg
16407787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16475796 Supplier Medchem Express Supplier No. HY12281
MedChemExpress FPH2 induces of functional proliferation of primary human hepatocytes and may lead to the development of new therapeutics for liver diseases.
FPH2 induces of functional proliferation of primary human hepatocytes and may lead to the development of new therapeutics for liver diseases.
Specifications
Chemical Name or Material FPH2
CAS 957485-64-2
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClN5O2S
Quantity 100 mg
Synonym BRD-9424
Solubility Information DMSO : 100 mg/mL (282.62 mM; Need ultrasonic)
SMILES O=C(C1=NN(CC)C=C1NC(NC2=CC(Cl)=CC=C2OC)=S)N
Molecular Weight (g/mol) 353.83
Formula Weight 353.83
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death