missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FOXP4 Polyclonal antibody apecifically detetects FOXP4 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications
Specifications
| Antigen | FOXP4 |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Concentration | 800 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | BC052803 |
| Gene Symbols | FOXP4 |
| Host Species | Rabbit |
| Immunogen | FOXP4 fusion protein Ag29953 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?