Learn More
Invitrogen™ eBioscience™ Foxp3 / Transcription Factor Fixation/Permeabilization Concentrate and Diluent
Product Code. 15151976
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Contains Sodium azide and formaldehyde
Product Code. 15151976
Brand: Invitrogen™ 00552100
Contains Sodium azide and formaldehyde
For staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines. Should be used with the Permeabilization Buffer. All three components sold together as the Foxp3 / Transcription Factor Staining Buffer Set.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Fixation Diluent, Fixation Reagent, Permeabilization Diluent, Permeabilization Reagent
Product Line eBioscience
Product Type Fixation/Permeabilization Diluent
For Research Use Only. Not for use in diagnostic procedures.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis