missing translation for 'onlineSavingsMsg'
Learn More

Proteintech FOXN1 Mouse anti-Human, Clone: 1D8B12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66337-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898115

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FOXN1 is a 648 amino acid protein belonging to the winged helix/forkhead family It contains 1 fork-head DNA-binding domain and is a transcriptional regulator involved in development. FOXN1 is involved in directing lineage choices of multi-potential progenitor epithelial cells and controls important regulating genes of lympho-epithelial interactions. It modulates keratinocyte differentiation in hair follicles and nail apparatus and its defects lead to T-cell immunodeficiency congenital alopecia, nail dystrophy, macroscopic nudity and an inborn dysgenesis of the thymus.
Specifications

Specifications

FOXN1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15353
FOXN1
FOXN1 Fusion Protein Ag18207
20 μL
Primary
Human
Antibody
IgG1
Western Blot
1D8B12
Unconjugated
FOXN1
FKHL20, forkhead box N1, Forkhead box protein N1, FOXN1, RONU, WHN
Mouse
Protein G
RUO
8456
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers