missing translation for 'onlineSavingsMsg'
Learn More

FOXH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20268-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889794

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to FOXH1.

FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
Specifications

Specifications

FOXH1
Polyclonal
Unconjugated
FOXH1
FAST 1, Fast 2, FAST1, FAST2, forkhead box H1, Forkhead box protein H1, FOXH1, hFAST 1
Rabbit
Antigen Affinity Chromatography
RUO
14106, 300054, 8928
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75593, O88621
FOXH1
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers