Description
Fowler Digital Micrometer features a thimble ratchet and lock.
- Provides instant readings to 0.00005 in. on the 6-digit LCD.
- Zero preset
- Hold
- Relative/absolute function
- Auto power on and off
- Carbide tipped anvils
- Includes 1.5 V battery, fitted storage case, and wrench for calibration.
- Range 0 to 1 in. (0 to 25 mm)
Specifications
Specifications
| Product Type | Ruler |
Product Title
¿Detecta una oportunidad de mejora?