Learn More
Labconco™ Four-Receptable Power Strip
Product Code. 11994809
Change view
Click to view available options
Amperage:
2 A
Includes:
4 receptacles
Product Type:
European Power Strip
Voltage:
250 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Amperage Includes Product Type Voltage unitSize
11994809 2 A 4 receptacles European Power Strip 250 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11994809 Supplier Labconco™ Supplier No. 5290400
Stock Up Your Lab
Use promo code 28007
View all promotions
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Provide electrical receptacles for connecting instruments and equipment
  • Electrical power extender for connecting equipment and instruments
  • North American electrical configuration
  • Adds four electrical receptacles
Specifications
Product Type European Power Strip
For Use With (Equipment) Electrical Equipment
Voltage 250 V
Amperage 2 A
For Use With (Application) To provide electrical receptacles for connecting instruments and equipment
Includes 4 receptacles
Weight (English) 1 lb.
Weight (Metric) 0.5 kg