Learn More
Thermo Scientific™ Four Inner-Door Option for Thermo Scientific™ Ultra-Low Temperature Freezers
Product Code. 11336361 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
HERAfreeze HFU300T, Revco UxF300
HERAfreeze HFU400T, Revco UxF400
HERAfreeze HFU500T, Revco UxF500
HERAfreeze HFU600T, Revco UxF600
HERAfreeze HFU700T, Revco UxF700
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11336361 HERAfreeze HFU500T, Revco UxF500 Each
11316361 HERAfreeze HFU300T, Revco UxF300 Each
11326361 HERAfreeze HFU400T, Revco UxF400 Each
11346361 HERAfreeze HFU600T, Revco UxF600 Each
11356361 HERAfreeze HFU700T, Revco UxF700 Each
5 options
Product Code. 11336361
Brand: Thermo Scientific™ 4D500
Only null left
Add to basket
Add to basket
Use with Thermo Scientific™ Revco™ UxF and HERAfreeze™ HFU T Series freezers.
Choose one of the Thermo Scientific™ Four Inner-Door Options Ultra-Low Temperature Freezers for your lab. Use these doors with Thermo Scientific™ Revco™ UxF and HERAfreeze™ HFU T Series freezers.
TRUSTED_SUSTAINABILITY
Specifications
Type Ultra-Low Freezer Inner-Door
No. of Doors 4
For Use With (Equipment) HERAfreeze HFU500T, Revco UxF500