Learn More
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser
Product Code. 11758286
Change view
Click to view available options
Capacity (Metric):
30 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11758286 50 mL Each
10272941 30 mL Each
2 options
Product Code. 11758286
Brand: Poulten & Graf GmbH 101 10648
Only null left
Add to basket
Add to basket
Poulten & Graf GmbH FORTUNA UNIVERSAL™ OPTIFIX™ HF Dispenser is for dispensing hydroflouric acid and all liquids that can be handled with the OPTIFIX™ dispensers.
  • No metal spring
  • No contamination of the dosing liquid
  • According to EN ISO 8655-5
  • Made of PTFE
Specifications
Quantity 1
Type Bottle Top Dispenser
Capacity (Metric) 50 mL