Learn More
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser
Product Code. 10314121
Change view
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
2.0 to 10 mL
20 to 100 mL
6 to 30 mL
Includes:
PTFE-coated Piston, Stopcock and Ventilator Valve
PTFE-encased Piston, Sealing Lip
Strongly PTFE-encased Special Piston
Unit Size:
Each
Product Code. Volume (Metric) Includes unitSize
10314121 1 to 5 mL PTFE-encased Piston, Sealing Lip Each
10252371 0.4 to 2 mL PTFE-encased Piston, Sealing Lip Each
10684321 2.0 to 10 mL PTFE-encased Piston, Sealing Lip Each
10654151 10 to 50 mL PTFE-encased Piston, Sealing Lip Each
10058720 20 to 100 mL PTFE-encased Piston, Sealing Lip Each
10549491 6 to 30 mL PTFE-encased Piston, Sealing Lip Each
10469652 2 to 10 mL Strongly PTFE-encased Special Piston Each
10719212 2 to 10 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11778286 6 to 30 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11788286 10 to 50 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11792358 1 to 5 mL Strongly PTFE-encased Special Piston Each
10341222 1 to 5 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
11768286 0.4 to 2 mL PTFE-coated Piston, Stopcock and Ventilator Valve Each
13 options
Product Code. 10314121
Brand: Poulten Graf™ 101 10733
Only null left
Add to basket
Add to basket
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With certificate, individually numbered
  • Glass dispensing cylinder is coated with plastic to provide breakage protection
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Product Type Safety Dispenser
Volume (Metric) 1 to 5 mL
Includes PTFE-encased Piston, Sealing Lip