Learn More
Emma Safety Footwear Fornax Safety Shoes
Product Code. 15998251
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15998251 39 1 pair
15908261 40 1 pair
15918261 41 1 pair
15928261 42 1 pair
15938261 43 1 pair
15948261 44 1 pair
15958261 45 1 pair
15968261 46 1 pair
15978261 47 1 pair
15988261 48 1 pair
15998261 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15998251 Supplier Emma Safety Footwear Supplier No. MM15884839
  • ONLINE EXCLUSIVE PRICE
    2189.25 NOK
    1281.00 NOK / 1 pairs
    Save 908.25 NOK
    41% Off
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Extra high boots for extra levels of protection with heat and cold insulated (HI and CI). Ankle protection with added padding provides a more comfortable feel and better fit. Fornax is very easy to put on and take off; the vertical eyelets on the shoe make lacing laces very simple.
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and cold insulated
  • 3M™ Reflective Striping
  • Full grain leather construction
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Fornax
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 39
Toe Material Steel