Learn More
Sartorius™ Formulation VI Q-App for Cubis™ MSA Balances
Shop All Sartorius Products
Click to view available options
:
Formulation V1
Description
Customize a Cubis MSA laboratory balance at any time by downloading specific applications, called Q-Apps, directly into the balance.
- Once installed, Q-Apps guide users step-by-step through a specific weighing workflow
- Integrating SOPs directly into the Cubis adapts the balance to a process
- Functions as embedded software on which fewer validation requirements are needed
- YAPPOF: Formulation VI – a simple formulation with integrated recipe memory
Specifications
Specifications
| For Use With (Equipment) | Cubis™ MSA Series Balance |
| For Use With (Application) | Simple formulation with integrated recipe memory |
| Product Line | Cubis™ Q-Apps |
| Type | Q-App License |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur