Learn More
Formanilide, 99+%
Product Code. 10296010 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
Product Code. 10296010
Brand: Thermo Scientific Acros 158421000
Request Bulk or Custom Format
CAS: 103-70-8 | C7H7NO | 121.139 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-70-8
MDL Number MFCD00003276
InChI Key DYDNPESBYVVLBO-UHFFFAOYSA-N
Synonym formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
PubChem CID 7671
ChEBI CHEBI:42416
IUPAC Name N-phenylformamide
SMILES C1=CC=C(C=C1)NC=O
Specifications
Melting Point 47°C to 49°C
Color Beige-Gray to White
Density 1.1400g/mL
Boiling Point 271°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Molecular Formula C7H7NO
Linear Formula HCONHC6H5
Quantity 100 g
Beilstein 12, 230
Merck Index 15, 4266
Specific Gravity 1.14
Solubility Information Solubility in water: 25.4g/L (20 c)
Molecular Weight (g/mol) 121.14
Formula Weight 121.14
Percent Purity 99+%
Physical Form Crystals
Chemical Name or Material Formanilide, 99+%