Learn More
Formamidine Acetate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 3473-63-0 |
| Melting Point | 156°C to 160°C |
| MDL Number | MFCD00012866 |
| Quantity | 100 g |
| Synonym | Formamidine acetate, Formamidine acetate |
| Molecular Weight (g/mol) | 44.06 (Free Base Basis) |
| Grade | Research |