missing translation for 'onlineSavingsMsg'
Learn More

Formamide, ≥99.0% (GC), Reagent Grade, Honeywell™

Product Code. 15627880 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
250 mL
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1 L
100mL
2.5 L
250mL
4 L
500mL
5mL
This item is not returnable. View return policy

Product Code. 15627880

Brand: Honeywell Research Chemicals F7503250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

pH 4.0 to 5.0 (at 20°C)
Melting Point 1°C to 3°C
Color Colorless
Density 1.134g/mL at 25°C
Boiling Point 210°C (1013 hPa)
Quantity 250 mL
Flash Point 175°C (347°F)
Ignition Point 932°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCONH2
Vapor Pressure 0.08mmHg (20°C), 30mmHg (129°C)
UN Number NONH for all modes of transport
Beilstein 505995
Solubility Information Fully miscible in water
Formula Weight 45.04g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Formamide
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.