Learn More
WTW™ Formaldehyde Cell Test Kit
With certificate for optimum precision.
Brand: WTW™ 250406
Additional Details : Weight : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For infrequent or very large sample throughput in the laboratory
- Sample adding, inserting, measuring and reading
- Reaction cuvette with barcode or method selection
Specifications
Chromotropic acid | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
Formaldehyde (HCHO) cell test kit |
For disinfectants, preservatives, applications for cosmetics and chipboard | |
Certificates | |
0.10 - 8.00mg/I HCHO | |
Photometric |