Learn More
WTW™ Formaldehyde Reagent Test Kit
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Specifications
| Analytes | Barium Sulfate |
| For Use With (Application) | For disinfectants, preservatives, applications for cosmetics and chipboard |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral photometers |
| Includes | Certificates |
| No. of Tests | 100 |
| Range | 0.02 - 8.00mg/I HCHO |
| Sufficient For | 100 Tests |
| Test Method | Photometric |
| Type | Sulfate (SO4) powder pillows test kit |