missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 4% Buffered, Solveco

Product Code. p-200087529
Click to view available options
Quantity:
1 L
5 L
25 L
Unit Size:
12 L
15 L
25 L
This item is not returnable. View return policy

Product Code. 18012193

Brand: Solveco Chemicals 1092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formaldehyde 4% (also known as Formalin 10%), saline-buffered for use in tissue sample fixation.

Formaldehyde 4% is a high-quality fixative solution specifically formulated for use in histopathology. This solution is ideal for preserving tissue specimens, ensuring their integrity and stability for accurate microscopic examination.

  • Effective Tissue Fixation: The 4% concentration of formaldehyde is optimized for halting enzymatic and bacterial degradation after tissue removal, preserving cellular and extracellular structures as closely as possible to their natural state.
  • Buffered Formulation: Buffered with salt, this solution maintains a stable pH level, which is crucial for preventing the formaldehyde from becoming acidic over time. This buffering ensures consistent fixation results and prevents potential degradation of the tissue sample.
  • Maintains Tissue Architecture: Cross-links proteins in the tissue, stabilizing cellular morphology and composition for reliable histological analysis.
  • Ease of Sectioning: Fixed tissues become firm and easy to cut into thin sections, facilitating smooth preparation for microscopic examination.
  • Long-term Storage: Provides excellent preservation, allowing tissue samples to be stored for extended periods without significant degradation.
  • Enhanced Staining: Ensures effective staining, making it easier to visualize and analyze different tissue components under the microscope.

Formaldehyde 4% is essential for accurate tissue fixation and reliable histopathological results, making it a staple in laboratory settings.

The product is CE-marked as a general IVD product in accordance with the In Vitro Diagnostic Directive (Regulation (EU) 2017/746 on in vitro diagnostic medical devices).

Chemical Identifiers

Molecular Weight (g/mol) 30.03

Specifications

Quantity 5 L
Percent Purity 4%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
compliance-icons compliance-icons
Product Identifier
  • Formaldehyde 4% Buffered
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.