Learn More
Thermo Scientific™ Forma™ Sample Probe Kits
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits.
Brand:  Thermo Scientific™ 195892
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11766649
  • 3626.65 NOK / Each
Estimated Shipment: 24-06-2024
Description
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits. Use probe kit with Forma 8600 Series Upright Freezers.
  • Useful for measuring sample temperature or top chamber temperature
  • Kits consist of probe and probe kit
Specifications
Specifications
Customer installed
Sample Probe
Upright